Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGTTGGCTCTGCTCCACCATCG[A/G]CCTTAACCAGCCCAGCACACCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176872 MIM: 180479 MIM: 605979 | ||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MAP2K1 - mitogen-activated protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002755.3 | 1487 | Intron | NP_002746.1 | |||
XM_011521783.2 | 1487 | Intron | XP_011520085.1 | |||
XM_017022411.1 | 1487 | Missense Mutation | GAC,GGC | D,G 354 | XP_016877900.1 | |
XM_017022412.1 | 1487 | Missense Mutation | GAC,GGC | D,G 332 | XP_016877901.1 | |
XM_017022413.1 | 1487 | Missense Mutation | GAC,GGC | D,G 204 | XP_016877902.1 |
MIR4512 - microRNA 4512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL4 - ribosomal protein L4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNAPC5 - small nuclear RNA activating complex polypeptide 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |