Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAACTCTTGAGGTAGAAATGGTA[A/G]ACCAGGAATAAAATTATTTTCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604875 MIM: 608659 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO9A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYO9A - myosin IXA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SENP8 - SUMO/sentrin peptidase family member, NEDD8 specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166340.1 | Intron | NP_001159812.1 | ||||
NM_001172109.1 | Intron | NP_001165580.1 | ||||
NM_001172110.1 | Intron | NP_001165581.1 | ||||
NM_001172111.1 | Intron | NP_001165582.1 | ||||
NM_145204.3 | Intron | NP_660205.3 | ||||
XM_005254157.3 | Intron | XP_005254214.1 | ||||
XM_011521215.2 | Intron | XP_011519517.1 | ||||
XM_011521217.2 | Intron | XP_011519519.1 |
Set Membership: |
HapMap |