Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTGAACAATATCTTTATTTAG[C/G]TCAGGTAGTTGTTCTTACACATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108330 | ||||||||||||||||||||
Literature Links: |
CYP1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP1A1 - cytochrome P450 family 1 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000499.4 | 2656 | UTR 3 | NP_000490.1 | |||
NM_001319216.1 | 2656 | UTR 3 | NP_001306145.1 | |||
NM_001319217.1 | 2656 | UTR 3 | NP_001306146.1 | |||
XM_017021953.1 | 2656 | UTR 3 | XP_016877442.1 |