Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGTCAGCACCCTCACAGAGCCA[A/G]CTAGGTACTGGGCCCAGGGGCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108330 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP1A1 - cytochrome P450 family 1 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000499.4 | 2086 | UTR 3 | NP_000490.1 | |||
NM_001319216.1 | 2086 | UTR 3 | NP_001306145.1 | |||
NM_001319217.1 | 2086 | UTR 3 | NP_001306146.1 | |||
XM_017021953.1 | 2086 | UTR 3 | XP_016877442.1 |