Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTTTAAATGGTGCTTTGGAAAA[A/T]TTTATTTTACTTAAAGTTCTGGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601149 | ||||||||||||||||||||
Literature Links: |
IDH3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IDH3A - isocitrate dehydrogenase 3 (NAD(+)) alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005530.2 | Intron | NP_005521.1 | ||||
XM_005254336.2 | Intron | XP_005254393.1 | ||||
XM_017022118.1 | Intron | XP_016877607.1 | ||||
XM_017022119.1 | Intron | XP_016877608.1 | ||||
XM_017022120.1 | Intron | XP_016877609.1 |