Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTATCCCCTTTCCCTCACTGGA[C/G]TTAGGTGGGTTTTTGCTATGATACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614681 MIM: 147582 | ||||||||||||||||||||
Literature Links: |
HYKK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HYKK - hydroxylysine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013619.3 | Intron | NP_001013641.2 | ||||
NM_001083612.1 | Intron | NP_001077081.1 | ||||
XM_011521231.2 | Intron | XP_011519533.1 | ||||
XM_017021922.1 | Intron | XP_016877411.1 |
IREB2 - iron responsive element binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |