Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGTTCTAATATTTGAGTTTTCT[C/G]TGAGGAAAAATAGTTGCAAGATTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610183 | ||||||||||||||||||||
Literature Links: |
ZFAND6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFAND6 - zinc finger AN1-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242911.1 | Intron | NP_001229840.1 | ||||
NM_001242912.1 | Intron | NP_001229841.1 | ||||
NM_001242913.1 | Intron | NP_001229842.1 | ||||
NM_001242914.1 | Intron | NP_001229843.1 | ||||
NM_001242915.1 | Intron | NP_001229844.1 | ||||
NM_001242916.1 | Intron | NP_001229845.1 | ||||
NM_001242917.1 | Intron | NP_001229846.1 | ||||
NM_001242918.1 | Intron | NP_001229847.1 | ||||
NM_001242919.1 | Intron | NP_001229848.1 | ||||
NM_019006.3 | Intron | NP_061879.2 | ||||
XM_011521684.2 | Intron | XP_011519986.1 |