Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTTAAAAAAAAAAAAAAAAAAA[A/G]ATCTTGGGTTCAATATGTCCTTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CFAP161 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP161 - cilia and flagella associated protein 161 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173528.2 | Intron | NP_775799.2 | ||||
XM_006720408.2 | Intron | XP_006720471.1 | ||||
XM_011521280.1 | Intron | XP_011519582.1 | ||||
XM_017021963.1 | Intron | XP_016877452.1 |