Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGACCTTACAGATACCTTAGAG[C/T]GTCTTTCTATTGCTTTAAATAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP161 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP161 - cilia and flagella associated protein 161 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173528.2 | Intron | NP_775799.2 | ||||
XM_006720408.2 | Intron | XP_006720471.1 | ||||
XM_011521280.1 | Intron | XP_011519582.1 | ||||
XM_017021963.1 | Intron | XP_016877452.1 |