Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGAATATAAGAAGAGAAAGCTGA[A/G]GAGAGGGGTAAGAGACCGATTTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613624 | ||||||||||||||||||||
Literature Links: |
ZNF592 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF592 - zinc finger protein 592 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014630.2 | Intron | NP_055445.2 | ||||
XM_005254996.3 | Intron | XP_005255053.1 | ||||
XM_011522246.2 | Intron | XP_011520548.1 | ||||
XM_011522247.2 | Intron | XP_011520549.1 | ||||
XM_011522248.2 | Intron | XP_011520550.1 | ||||
XM_017022734.1 | Intron | XP_016878223.1 |