Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGTCACTTGCTATCCGTGGGAC[C/G]TTATGCAAATAAATCTGAGCCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611851 MIM: 611977 | ||||||||||||||||||||
Literature Links: |
MRPL46 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL46 - mitochondrial ribosomal protein L46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022163.3 | 559 | Intron | NP_071446.2 |
MRPS11 - mitochondrial ribosomal protein S11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321970.1 | 559 | Intron | NP_001308899.1 | |||
NM_001321972.1 | 559 | UTR 5 | NP_001308901.1 | |||
NM_001321973.1 | 559 | Intron | NP_001308902.1 | |||
NM_001321974.1 | 559 | Intron | NP_001308903.1 | |||
NM_001321976.1 | 559 | UTR 5 | NP_001308905.1 | |||
NM_022839.4 | 559 | Intron | NP_073750.2 | |||
NM_176805.3 | 559 | Intron | NP_789775.1 |