Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGAAGATAAGTCACATAATCA[A/G]GTTTGTGTTTCAAAAACATCACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCI - Fanconi anemia complementation group I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113378.1 | Intron | NP_001106849.1 | ||||
NM_018193.2 | Intron | NP_060663.2 | ||||
XM_011521756.2 | Intron | XP_011520058.1 | ||||
XM_011521757.2 | Intron | XP_011520059.1 | ||||
XM_011521764.2 | Intron | XP_011520066.1 | ||||
XM_011521767.2 | Intron | XP_011520069.1 |