Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTAGCCCTGCTCTGGCGGCCCG[G/T]CCCGGAGCTGGAGGCCGCTCGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BLM - Bloom syndrome RecQ like helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000057.3 | Intron | NP_000048.1 | ||||
NM_001287246.1 | Intron | NP_001274175.1 | ||||
NM_001287247.1 | Intron | NP_001274176.1 | ||||
NM_001287248.1 | Intron | NP_001274177.1 | ||||
XM_006720632.2 | Intron | XP_006720695.1 | ||||
XM_011521881.2 | Intron | XP_011520183.1 | ||||
XM_011521882.2 | Intron | XP_011520184.1 |
CRTC3-AS1 - CRTC3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |