Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCTGCATTGATAGGAGATGAAT[A/G]TTCGGAAGGTGGGACCCCAGTTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSM2B - acyl-CoA synthetase medium-chain family member 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105069.1 | Intron | NP_001098539.1 | ||||
NM_182617.3 | Intron | NP_872423.3 | ||||
XM_005255304.3 | Intron | XP_005255361.1 | ||||
XM_011545824.2 | Intron | XP_011544126.1 | ||||
XM_017023205.1 | Intron | XP_016878694.1 |