Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 145505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSM3 - acyl-CoA synthetase medium-chain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005622.3 | 483 | Missense Mutation | GGA,GTA | G,V 101 | NP_005613.2 | |
NM_202000.2 | 483 | Missense Mutation | GGA,GTA | G,V 101 | NP_973729.1 | |
XM_011545911.1 | 483 | Missense Mutation | GGA,GTA | G,V 101 | XP_011544213.1 | |
XM_017023523.1 | 483 | Missense Mutation | GGA,GTA | G,V 101 | XP_016879012.1 | |
XM_017023524.1 | 483 | Missense Mutation | GGA,GTA | G,V 101 | XP_016879013.1 |
ERI2 - ERI1 exoribonuclease family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |