Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATGCATCGCAAGACATGCTCCC[C/T]CCTTCCTCCCCGCTGGGCTCCAGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612897 MIM: 613620 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC107983970 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC107983970 - uncharacterized LOC107983970 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYLPF - myosin light chain, phosphorylatable, fast skeletal muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324458.1 | Intron | NP_001311387.1 | ||||
NM_001324459.1 | Intron | NP_001311388.1 | ||||
NM_013292.4 | Intron | NP_037424.2 |
SEPT1 - septin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10B - TBC1 domain family member 10B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF48 - zinc finger protein 48 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |