Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGGGTCCCATGCTGCAGGGGC[A/G]CCCTGGTTTTCAGGGTAAGGAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGAL - integrin subunit alpha L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114380.1 | 607 | Intron | NP_001107852.1 | |||
NM_002209.2 | 607 | Missense Mutation | CAC,CGC | H,R 144 | NP_002200.2 | |
XM_005255313.1 | 607 | Missense Mutation | CAC,CGC | H,R 144 | XP_005255370.1 | |
XM_006721044.1 | 607 | Intron | XP_006721107.1 | |||
XM_011545849.1 | 607 | Missense Mutation | CAC,CGC | H,R 48 | XP_011544151.1 |