Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTCCACACACACAAAAAAAAAA[A/T]TTAAAAATTAGCTAGGTATGGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603795 | ||||||||||||||||||||
Literature Links: |
LITAF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LITAF - lipopolysaccharide induced TNF factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136472.1 | Intron | NP_001129944.1 | ||||
NM_001136473.1 | Intron | NP_001129945.1 | ||||
NM_004862.3 | Intron | NP_004853.2 | ||||
XM_006720982.2 | Intron | XP_006721045.1 | ||||
XM_006720983.3 | Intron | XP_006721046.1 | ||||
XM_006720984.3 | Intron | XP_006721047.1 | ||||
XM_006720985.3 | Intron | XP_006721048.1 | ||||
XM_011522754.2 | Intron | XP_011521056.1 | ||||
XM_017023896.1 | Intron | XP_016879385.1 |