Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTCAGAAGGAAAGAAGGTCTT[C/G]TAAGTGCTCTCAATGTGATAGGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C16orf45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C16orf45 - chromosome 16 open reading frame 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142469.1 | Intron | NP_001135941.1 | ||||
NM_033201.2 | Intron | NP_149978.1 | ||||
XM_011522731.2 | Intron | XP_011521033.1 | ||||
XM_017023824.1 | Intron | XP_016879313.1 |