Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATCCAAAACTTAGCAAGAATG[C/T]ACTATTACCTACACTTTCTAAAGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602212 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507577 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100507577 - uncharacterized LOC100507577 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIAH1 - siah E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006610.1 | Intron | NP_001006611.1 | ||||
NM_003031.3 | Intron | NP_003022.3 | ||||
XM_006721246.1 | Intron | XP_006721309.1 | ||||
XM_011523279.1 | Intron | XP_011521581.1 |
Set Membership: |
HapMap |