Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTGTCCGTCAGCCGGTCTACA[A/G]CACCCCCTGTTTTTGCTTTCTGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616957 | ||||||||||||||||||||
Literature Links: |
LRRC36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC36 - leucine rich repeat containing 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPPP3 - tubulin polymerization promoting protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015964.3 | 524 | Missense Mutation | GCT,GTT | A,V 121 | NP_057048.2 | |
NM_016140.3 | 524 | Missense Mutation | GCT,GTT | A,V 121 | NP_057224.2 |
ZDHHC1 - zinc finger DHHC-type containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |