Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCTATGGAAATTTTCTGTAACC[A/C]CCACATTTAATGTAAGAACATGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604167 | ||||||||||||||||||||
Literature Links: |
CTCF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTCF - CCCTC-binding factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191022.1 | Intron | NP_001177951.1 | ||||
NM_006565.3 | Intron | NP_006556.1 | ||||
XM_005255775.3 | Intron | XP_005255832.1 | ||||
XM_017022868.1 | Intron | XP_016878357.1 | ||||
XM_017022869.1 | Intron | XP_016878358.1 |