Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCTGCCTCTCACTAATTTTGAT[G/T]GTGATGCTAGAATTCAGCTTTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 192090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH1 - cadherin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317184.1 | Intron | NP_001304113.1 | ||||
NM_001317185.1 | Intron | NP_001304114.1 | ||||
NM_001317186.1 | Intron | NP_001304115.1 | ||||
NM_004360.4 | Intron | NP_004351.1 | ||||
XM_011523488.1 | Intron | XP_011521790.1 | ||||
XM_011523489.1 | Intron | XP_011521791.1 |