Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTTTCTTCAACCCCATTAGAGC[C/T]GTTCCACTTGCCTTTGGGTACTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602027 | ||||||||||||||||||||
Literature Links: |
TERF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TERF2 - telomeric repeat binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005652.4 | 1405 | Missense Mutation | AGC,GGC | S,G 455 | NP_005643.2 | |
XM_005256121.3 | 1405 | Missense Mutation | AGC,GGC | S,G 458 | XP_005256178.1 | |
XM_005256122.3 | 1405 | Missense Mutation | AGC,GGC | S,G 458 | XP_005256179.1 | |
XM_005256123.3 | 1405 | Missense Mutation | AGC,GGC | S,G 455 | XP_005256180.1 | |
XM_005256124.3 | 1405 | Missense Mutation | AGC,GGC | S,G 377 | XP_005256181.1 |
TMED6 - transmembrane p24 trafficking protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |