Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAAGTGCCTGCTCCTGCAGCT[C/T]TGAGTGCATATGCGCTCCCCACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUK - fucokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145059.2 | Intron | NP_659496.2 | ||||
XM_006721162.1 | Intron | XP_006721225.1 | ||||
XM_011522928.1 | Intron | XP_011521230.1 | ||||
XM_011522929.1 | Intron | XP_011521231.1 | ||||
XM_011522930.2 | Intron | XP_011521232.1 | ||||
XM_011522931.2 | Intron | XP_011521233.1 | ||||
XM_017023012.1 | Intron | XP_016878501.1 | ||||
XM_017023013.1 | Intron | XP_016878502.1 | ||||
XM_017023014.1 | Intron | XP_016878503.1 |