Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGGTAATTATGTGAGGATATG[C/T]GGTTTCTAGCCTGTTTCCATTCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614951 | ||||||||||||||||||||
Literature Links: |
HEATR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEATR3 - HEAT repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256013.2 | Intron | XP_005256070.1 | ||||
XM_011523188.2 | Intron | XP_011521490.1 | ||||
XM_017023385.1 | Intron | XP_016878874.1 | ||||
XM_017023386.1 | Intron | XP_016878875.1 |