Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCACGCCCGCCGCCCCTCCCGC[A/G]TCCTGGCCCCTTCCCCACCCGACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC16A5 - solute carrier family 16 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271765.1 | 125 | UTR 5 | NP_001258694.1 | |||
NM_004695.3 | 125 | Intron | NP_004686.1 | |||
XM_005257790.1 | 125 | Intron | XP_005257847.1 | |||
XM_011525462.1 | 125 | Intron | XP_011523764.1 | |||
XM_017025293.1 | 125 | Intron | XP_016880782.1 |