Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCCTGAGTCAGCCTGCTTTCT[C/T]TCTCAGGTGAGGCGGAACCAACCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600864 MIM: 603877 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1D PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CSNK1D - casein kinase 1 delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6787 - microRNA 6787 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A3 - solute carrier family 16 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042422.2 | Intron | NP_001035887.1 | ||||
NM_001042423.2 | Intron | NP_001035888.1 | ||||
NM_001206950.1 | Intron | NP_001193879.1 | ||||
NM_001206951.1 | Intron | NP_001193880.1 | ||||
NM_001206952.1 | Intron | NP_001193881.1 | ||||
NM_004207.3 | Intron | NP_004198.1 | ||||
XM_011523607.1 | Intron | XP_011521909.1 |
Set Membership: |
HapMap |