Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCAGGTGCCACCACACCTGGCC[A/G]CTTTTTTTTCTTAAACCGATTTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616864 | ||||||||||||||||||||
Literature Links: |
C17orf62 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C17orf62 - chromosome 17 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HEXDC - hexosaminidase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173620.2 | Intron | NP_775891.2 | ||||
XM_005256346.3 | Intron | XP_005256403.1 | ||||
XM_005256347.4 | Intron | XP_005256404.1 | ||||
XM_006722280.3 | Intron | XP_006722343.1 | ||||
XM_011523560.2 | Intron | XP_011521862.1 | ||||
XM_011523561.2 | Intron | XP_011521863.1 | ||||
XM_017024484.1 | Intron | XP_016879973.1 | ||||
XM_017024485.1 | Intron | XP_016879974.1 |