Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160770 | ||||||||||||||||||||
Literature Links: |
MYL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYL4 - myosin light chain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002841.1 | 39 | Intron | NP_001002841.1 | |||
NM_002476.2 | 39 | UTR 5 | NP_002467.1 | |||
XM_005257391.4 | 39 | Intron | XP_005257448.1 | |||
XM_011524839.2 | 39 | Intron | XP_011523141.2 | |||
XM_017024683.1 | 39 | Intron | XP_016880172.1 | |||
XM_017024684.1 | 39 | UTR 5 | XP_016880173.1 |