Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATAATAATAAAAAAAGAAGCAAT[C/T]GGGATAGACAAGAAGGAGCAAGTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606779 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
EFCAB5 - EF-hand calcium binding domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145053.1 | Intron | NP_001138525.1 | ||||
NM_198529.3 | Intron | NP_940931.2 | ||||
XM_011524759.1 | Intron | XP_011523061.1 | ||||
XM_011524763.2 | Intron | XP_011523065.1 | ||||
XM_011524765.1 | Intron | XP_011523067.1 | ||||
XM_011524766.2 | Intron | XP_011523068.1 | ||||
XM_011524768.2 | Intron | XP_011523070.1 | ||||
XM_017024591.1 | Intron | XP_016880080.1 | ||||
XM_017024592.1 | Intron | XP_016880081.1 |
SSH2 - slingshot protein phosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |