Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTTAGCAACCAATGATAGCCAA[A/G]TACACTAGAAGAGCACTAACAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFCAB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EFCAB5 - EF-hand calcium binding domain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145053.1 | Intron | NP_001138525.1 | ||||
NM_198529.3 | Intron | NP_940931.2 | ||||
XM_011524759.1 | Intron | XP_011523061.1 | ||||
XM_011524763.2 | Intron | XP_011523065.1 | ||||
XM_011524765.1 | Intron | XP_011523067.1 | ||||
XM_011524766.2 | Intron | XP_011523068.1 | ||||
XM_011524768.2 | Intron | XP_011523070.1 | ||||
XM_017024591.1 | Intron | XP_016880080.1 | ||||
XM_017024592.1 | Intron | XP_016880081.1 |