Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAAGTATGCACTAAAGTATACCT[A/G]GAATGATGAAGGTTACCAGGATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601758 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371743 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371743 - uncharacterized LOC105371743 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX12 - peroxisomal biogenesis factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000286.2 | Intron | NP_000277.1 |
SNORD7 - small nucleolar RNA, C/D box 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |