Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCTATATCACCATTGTCCGCC[C/T]TTGAGCCTGGAAGGAAAATGAGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601025 MIM: 601758 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2B1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP2B1 - adaptor related protein complex 2 beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030006.1 | 72 | Intron | NP_001025177.1 | |||
NM_001282.2 | 72 | Intron | NP_001273.1 | |||
XM_005257937.3 | 72 | Intron | XP_005257994.1 | |||
XM_005257938.2 | 72 | Intron | XP_005257995.1 | |||
XM_005257941.2 | 72 | Intron | XP_005257998.1 | |||
XM_011524448.1 | 72 | UTR 5 | XP_011522750.1 | |||
XM_011524449.2 | 72 | Intron | XP_011522751.1 | |||
XM_011524450.1 | 72 | Intron | XP_011522752.1 | |||
XM_011524451.1 | 72 | Intron | XP_011522753.1 | |||
XM_011524452.1 | 72 | Intron | XP_011522754.1 | |||
XM_011524453.1 | 72 | Intron | XP_011522755.1 | |||
XM_011524454.1 | 72 | Intron | XP_011522756.1 | |||
XM_011524455.2 | 72 | Intron | XP_011522757.1 | |||
XM_017024284.1 | 72 | Intron | XP_016879773.1 | |||
XM_017024285.1 | 72 | Intron | XP_016879774.1 | |||
XM_017024286.1 | 72 | Intron | XP_016879775.1 | |||
XM_017024287.1 | 72 | Intron | XP_016879776.1 |
PEX12 - peroxisomal biogenesis factor 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |