Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAGACATTTGACACATAGTAG[A/G]CATTTCCTATATAGTAGGGTTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL5 - C-C motif chemokine ligand 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278736.1 | Intron | NP_001265665.1 | ||||
NM_002985.2 | Intron | NP_002976.2 |
HEATR9 - HEAT repeat containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |