Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGGAGCAGTTAGAAATGCAGA[C/T]TCTTCCCTACTCCAGACTACGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609194 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABLES1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABLES1 - Cdk5 and Abl enzyme substrate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100619.2 | Intron | NP_001094089.1 | ||||
NM_001256438.1 | Intron | NP_001243367.1 | ||||
NM_138375.2 | Intron | NP_612384.1 |
LOC105372022 - uncharacterized LOC105372022 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |