Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTGCGAGACAGGGTCTCACTT[G/T]GTGGCTCAGGCTGGAGTGTACTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607461 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DYM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DYM - dymeclin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017653.3 | 3074 | Intron | NP_060123.3 | |||
XM_006722488.2 | 3074 | UTR 3 | XP_006722551.1 | |||
XM_006722490.2 | 3074 | Intron | XP_006722553.1 | |||
XM_006722491.2 | 3074 | Intron | XP_006722554.1 | |||
XM_006722492.3 | 3074 | Intron | XP_006722555.1 | |||
XM_011526036.1 | 3074 | UTR 3 | XP_011524338.1 | |||
XM_011526037.1 | 3074 | UTR 3 | XP_011524339.1 | |||
XM_011526038.1 | 3074 | UTR 3 | XP_011524340.1 | |||
XM_011526039.1 | 3074 | Intron | XP_011524341.1 | |||
XM_011526040.1 | 3074 | UTR 3 | XP_011524342.1 | |||
XM_011526041.1 | 3074 | UTR 3 | XP_011524343.1 | |||
XM_011526042.1 | 3074 | Intron | XP_011524344.1 | |||
XM_017025795.1 | 3074 | UTR 3 | XP_016881284.1 | |||
XM_017025796.1 | 3074 | UTR 3 | XP_016881285.1 | |||
XM_017025797.1 | 3074 | Intron | XP_016881286.1 | |||
XM_017025798.1 | 3074 | Intron | XP_016881287.1 | |||
XM_017025799.1 | 3074 | Intron | XP_016881288.1 | |||
XM_017025800.1 | 3074 | UTR 3 | XP_016881289.1 | |||
XM_017025801.1 | 3074 | Intron | XP_016881290.1 |
MIR4744 - microRNA 4744 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |