Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGAGGCTTAATTGGCTCACAGTT[C/T]TTCAGGCTGTAGAGGCTTCTGCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114020 | ||||||||||||||||||||
Literature Links: |
CDH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH2 - cadherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308176.1 | Intron | NP_001295105.1 | ||||
NM_001792.4 | Intron | NP_001783.2 | ||||
XM_011525788.1 | Intron | XP_011524090.1 | ||||
XM_017025514.1 | Intron | XP_016881003.1 |