Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATGTGATTTTAGGCAACTTACAT[A/G]TATACTTTTTGAACCTTAGTTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH2 - cadherin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308176.1 | Intron | NP_001295105.1 | ||||
NM_001792.4 | Intron | NP_001783.2 | ||||
XM_011525788.1 | Intron | XP_011524090.1 | ||||
XM_017025514.1 | Intron | XP_016881003.1 |