Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTTATTACATAACATTTAATG[C/T]ATACAAGTGAATAAATGAGTAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIB1 - mindbomb E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020774.3 | Intron | NP_065825.1 | ||||
XM_011526098.1 | Intron | XP_011524400.1 | ||||
XM_017025873.1 | Intron | XP_016881362.1 | ||||
XM_017025874.1 | Intron | XP_016881363.1 | ||||
XM_017025875.1 | Intron | XP_016881364.1 |