Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTATTCCTGGCCCTGTGTGAGTAAC[A/G]AGCACTTTTCTGAAATCCTTTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604495 MIM: 604480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCER2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCER2 - coiled-coil glutamate rich protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFKBIB - NFKB inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243116.1 | Intron | NP_001230045.1 | ||||
NM_002503.4 | Intron | NP_002494.2 | ||||
XM_006723226.2 | Intron | XP_006723289.2 | ||||
XM_006723227.3 | Intron | XP_006723290.3 |
SIRT2 - sirtuin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |