Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGGAGCTGATCTCCTCAGTC[C/T]CCCTGCTTTTCCCCGGTCTGCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACP7 - acid phosphatase 7, tartrate resistant (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004318.2 | 235 | UTR 5 | NP_001004318.2 | |||
XM_011526966.2 | 235 | UTR 5 | XP_011525268.1 | |||
XM_011526967.2 | 235 | UTR 5 | XP_011525269.1 | |||
XM_011526969.1 | 235 | UTR 5 | XP_011525271.1 | |||
XM_017026807.1 | 235 | UTR 5 | XP_016882296.1 | |||
XM_017026808.1 | 235 | UTR 5 | XP_016882297.1 |