Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTAACACAACAGAATGTATGGCC[A/G]GGCGCAGTGGCTCCTGCCTGTAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615698 | ||||||||||||||||||||
Literature Links: |
MIR6796 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6796 - microRNA 6796 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLD3 - phospholipase D family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031696.3 | Intron | NP_001026866.1 | ||||
NM_001291311.1 | Intron | NP_001278240.1 | ||||
NM_012268.3 | Intron | NP_036400.2 | ||||
XM_005258704.1 | Intron | XP_005258761.1 | ||||
XM_005258707.4 | Intron | XP_005258764.1 | ||||
XM_005258708.3 | Intron | XP_005258765.1 | ||||
XM_005258709.4 | Intron | XP_005258766.1 | ||||
XM_005258710.4 | Intron | XP_005258767.1 | ||||
XM_006723122.1 | Intron | XP_006723185.1 | ||||
XM_011526692.1 | Intron | XP_011524994.1 | ||||
XM_011526693.1 | Intron | XP_011524995.1 | ||||
XM_017026546.1 | Intron | XP_016882035.1 | ||||
XM_017026547.1 | Intron | XP_016882036.1 | ||||
XM_017026548.1 | Intron | XP_016882037.1 | ||||
XM_017026549.1 | Intron | XP_016882038.1 |