Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTGTGCTGGGAGCTCTCCATTA[A/G]CTAGAGCTCCTGGTTGGGGTGGAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123310 MIM: 606495 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CKM - creatine kinase, M-type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001824.4 | 1579 | UTR 3 | NP_001815.2 | |||
XM_017026240.1 | 1579 | UTR 3 | XP_016881729.1 |
MARK4 - microtubule affinity regulating kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |