Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCTGATTTGGCAAAAGGAAGTG[A/G]TTTCCTGTCCCCAGTGATTTCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126340 MIM: 607463 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC2 - ERCC excision repair 2, TFIIH core complex helicase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R13L - protein phosphatase 1 regulatory subunit 13 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142502.1 | 2924 | UTR 3 | NP_001135974.1 | |||
NM_006663.3 | 2924 | UTR 3 | NP_006654.2 | |||
XM_017026177.1 | 2924 | UTR 3 | XP_016881666.1 | |||
XM_017026178.1 | 2924 | UTR 3 | XP_016881667.1 | |||
XM_017026179.1 | 2924 | Intron | XP_016881668.1 |