Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGCAGCGGAGATTAGAAGATA[C/T]TCTTGAAGGCTTCCGGGACGGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114183 MIM: 600022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CALM3 - calmodulin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP5D1 - PPP5 tetratricopeptide repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205281.1 | Intron | NP_001192210.1 | ||||
XM_005258397.3 | Intron | XP_005258454.1 | ||||
XM_006722960.3 | Intron | XP_006723023.1 | ||||
XM_017026125.1 | Intron | XP_016881614.1 |
PTGIR - prostaglandin I2 (prostacyclin) receptor (IP) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |