Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTCCCCGCTTTGGGTGTTGTGA[C/T]GCTTCAATGAGTGAAGACTTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLTSCR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLTSCR1 - glioma tumor suppressor candidate region gene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015711.3 | Intron | NP_056526.3 | ||||
XM_005258833.4 | Intron | XP_005258890.1 | ||||
XM_006723180.3 | Intron | XP_006723243.1 | ||||
XM_011526882.2 | Intron | XP_011525184.1 | ||||
XM_011526883.2 | Intron | XP_011525185.1 |