Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCAGGAAACTGAGGCTCCAAG[A/C]GATGAGTGATTTGCCCCAGGCCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNGR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SYNGR4 - synaptogyrin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012451.3 | 176 | Intron | NP_036583.2 | |||
XM_005258693.2 | 176 | Intron | XP_005258750.1 | |||
XM_005258694.2 | 176 | Intron | XP_005258751.1 | |||
XM_011526686.2 | 176 | UTR 5 | XP_011524988.1 | |||
XM_017026541.1 | 176 | Intron | XP_016882030.1 |
TMEM143 - transmembrane protein 143 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |