Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTCTATGGAGCTTTCGCCTACC[C/T]GGAGAGGCCCAGAAAATTGACCGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602488 MIM: 603953 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTH2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYTH2 - cytohesin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004228.6 | 762 | Silent Mutation | CCC,CCT | P,P 154 | NP_004219.3 | |
NM_017457.5 | 762 | Silent Mutation | CCC,CCT | P,P 154 | NP_059431.1 | |
XM_006723472.2 | 762 | Silent Mutation | CCC,CCT | P,P 176 | XP_006723535.1 | |
XM_006723473.2 | 762 | Silent Mutation | CCC,CCT | P,P 176 | XP_006723536.1 |
KCNJ14 - potassium voltage-gated channel subfamily J member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372430 - uncharacterized LOC105372430 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |