Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACATTTACCGCCTGTGTATATG[C/T]GCCAGACACTGTTTTAATCCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182100 MIM: 610349 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUT2 - fucosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000511.5 | Intron | NP_000502.4 | ||||
NM_001097638.2 | Intron | NP_001091107.1 |
LOC105447645 - uncharacterized LOC105447645 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAMSTR - MEF2 activating motif and SAP domain containing transcriptional regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |